siRNA Details
| Gene Symbol: Isoc2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101598.1 | NP_001095068.1 | 3 | 399 | ||
| GenBank mRNA | BC158139.1 | - | 3 | 299 | 9007402 | |
| BG404719.1 | - | 3 | 398 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Isoc2a| Gene Symbol: Isoc2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101598.1 | NP_001095068.1 | 3 | 399 | ||
| GenBank mRNA | BC158139.1 | - | 3 | 299 | 9007402 | |
| BG404719.1 | - | 3 | 398 | |||