siRNA Details
| Gene Symbol: Pgpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023217.4 | NP_075706.1 | 5 | 1147 | ||
| GenBank mRNA | AJ278829.1 | - | 5 | 1079 | ||
| AK012658.1 | - | 5 | 1079 | |||
| AK047835.1 | - | 5 | 1091 | |||
| AK049632.1 | - | 5 | 1070 | |||
| BC043041.1 | - | 5 | 1057 | |||
| BC051938.1 | - | 5 | 1076 | 5717365 | ||

Pgpep1
Genomic Map