siRNA Details
| Gene Symbol: Rhox3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040089.3 | NP_001035178.3 | 1 | 64 | ||
| Gene Symbol: Rhox3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114157.1 | NP_001107629.1 | 3 | 284 | ||

Rhox3f
Genomic Map