siRNA Details
| Gene Symbol: Metap1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025633.3 | NP_079909.1 | 3 | 277 | ||
| GenBank mRNA | AK005241.1 | - | 3 | 274 | ||
| AK010067.1 | - | 3 | 277 | |||
| BC060739.1 | - | 3 | 264 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Metap1d| Gene Symbol: Metap1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025633.3 | NP_079909.1 | 3 | 277 | ||
| GenBank mRNA | AK005241.1 | - | 3 | 274 | ||
| AK010067.1 | - | 3 | 277 | |||
| BC060739.1 | - | 3 | 264 | |||