siRNA Details
| Gene Symbol: 1700030N03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006563.1 | - | 197 | |||
| AK007025.1 | - | 3 | 202 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700030N03Rik| Gene Symbol: 1700030N03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006563.1 | - | 197 | |||
| AK007025.1 | - | 3 | 202 | |||