siRNA Details
| Gene Symbol: Gm7861 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177526.1 | NP_001170997.1 | 2 | 226 | ||
| Gene Symbol: Gm7849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177518.1 | NP_001170989.1 | 2 | 226 | ||

Gm7861
Genomic Map