siRNA Details
| Gene Symbol: 1700017N19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081246.1 | NP_001074715.1 | 4 | 389 | ||
| GenBank mRNA | AK006074.1 | - | 3 | 360 | ||
| AK014917.1 | - | 4 | 367 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700017N19Rik| Gene Symbol: 1700017N19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081246.1 | NP_001074715.1 | 4 | 389 | ||
| GenBank mRNA | AK006074.1 | - | 3 | 360 | ||
| AK014917.1 | - | 4 | 367 | |||