siRNA Details
| Gene Symbol: 2410127L17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026120.4 | NP_080396.2 | 8 | 1683 | ||
| GenBank mRNA | AK004202.1 | - | 1 | 310 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2410127L17Rik| Gene Symbol: 2410127L17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026120.4 | NP_080396.2 | 8 | 1683 | ||
| GenBank mRNA | AK004202.1 | - | 1 | 310 | ||