siRNA Details
| Gene Symbol: Gskip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178613.3 | NP_848728.2 | 2 | 226 | ||
| GenBank mRNA | AK017049.1 | - | 750 | |||
| AK020785.1 | - | 2 | 164 | |||
| AK029484.1 | - | 2 | 138 | |||
| AK042664.1 | - | 2 | 187 | |||
| AK079775.1 | - | 2 | 172 | |||
| AK168962.1 | - | 2 | 190 | |||
| AK171097.1 | - | 2 | 203 | |||
| BC031494.1 | - | 2 | 194 | 3156862 | ||

Gskip
Genomic Map