siRNA Details
| Gene Symbol: Bcl2l14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025778.3 | NP_080054.1 | 3 | 351 | ||
| GenBank mRNA | AK008682.1 | - | 2 | 276 | ||
| AK016670.1 | - | 3 | 352 | |||
| AK016997.1 | - | 3 | 351 | |||
| AK018579.1 | - | 2 | 318 | |||
| AK035707.1 | - | 2 | 283 | |||
| AK160247.1 | - | 3 | 354 | |||
| BC025541.1 | - | 2 | 271 | 5253808 | ||

Bcl2l14
Genomic Map