siRNA Details
| Gene Symbol: Scgb2b12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281508.1 | NP_001268437.1 | 2 | 104 | ||
| GenBank mRNA | CK616067.1 | - | 2 | 112 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgb2b12| Gene Symbol: Scgb2b12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281508.1 | NP_001268437.1 | 2 | 104 | ||
| GenBank mRNA | CK616067.1 | - | 2 | 112 | ||