siRNA Details
| Gene Symbol: Plin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025836.3 | NP_080112.1 | 3, 4 | 346 | ||
| GenBank mRNA | AK004970.1 | - | 3, 4 | 347 | ||
| AK012517.1 | - | 3, 4 | 347 | |||
| AK049537.1 | - | 3, 4 | 347 | |||
| AK050140.1 | - | 3, 4 | 348 | |||
| AK132313.1 | - | 347 | ||||
| AK167214.1 | - | 3, 4 | 345 | |||
| AK197135.1 | - | 1, 2 | 104 | |||
| BC011116.1 | - | 3, 4 | 342 | 4023354 | ||
| DQ257436.1 | - | 3, 4 | 305 | |||

Plin3
Genomic Map