siRNA Details
| Gene Symbol: Tomm34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291155.1 | NP_001278084.1 | 6, 7 | 964 | ||
| NM_025996.5 | NP_080272.1 | 6, 7 | 1343 | |||
| GenBank mRNA | AK011786.1 | - | 6, 7 | 1343 | ||
| AK017699.1 | - | 6, 7 | 909 | |||
| AK075873.1 | - | 6, 7 | 961 | |||
| AK144450.1 | - | 3, 4 | 1131 | |||
| AK168329.1 | - | 6, 7 | 898 | |||
| BC018278.1 | - | 6, 7 | 894 | 4187356 | ||

Tomm34
Genomic Map