siRNA Details
| Gene Symbol: Cers4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026058.4 | NP_080334.3 | 8, 9 | 930 | ||
| GenBank mRNA | AK013554.1 | - | 8, 9 | 882 | ||
| AK021316.1 | - | 1, 2 | 37 | |||
| AK028937.1 | - | 8, 9 | 856 | |||
| AK028945.1 | - | 8, 9 | 833 | |||
| AK030280.1 | - | 8, 9 | 870 | |||
| AK134887.1 | - | 7, 8 | 1589 | |||
| AK177826.1 | - | 1, 2 | 56 | |||
| AY029531.1 | - | 8, 9 | 831 | |||
| BC003946.1 | - | 8, 9 | 817 | 3487343 | ||

Cers4
Genomic Map