siRNA Details
| Gene Symbol: Med4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026119.3 | NP_080395.1 | 3 | 255 | ||
| GenBank mRNA | AK010270.1 | - | 3 | 257 | ||
| AK010686.1 | - | 3 | 257 | |||
| AK078288.1 | - | 3 | 267 | |||
| AK145636.1 | - | 3 | 280 | |||
| BC024950.1 | - | 3 | 255 | 3992287 | ||

Med4
Genomic Map