siRNA Details
| Gene Symbol: Glipr1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026223.2 | NP_080499.1 | 1 | 164 | ||
| GenBank mRNA | AK014846.1 | - | 1 | 165 | ||
| AK015834.1 | - | 1 | 148 | |||
| AK161292.1 | - | 1 | 165 | |||
| BC118017.1 | - | 1 | 165 | 40102576 | ||

Glipr1l2
Genomic Map