siRNA Details
| Gene Symbol: Larp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026235.4 | NP_080511.2 | 3 | 775 | ||
| GenBank mRNA | AK017372.1 | - | 3 | 780 | ||
| AK020966.1 | - | 3 | 712 | |||
| AK035809.1 | - | 352 | ||||
| AK039614.1 | - | 3 | 758 | |||
| AK042616.1 | - | 3 | 707 | |||
| AK047452.1 | - | 1 | 79 | |||
| AK087521.1 | - | 3 | 778 | |||
| BC090615.1 | - | 3 | 907 | 6809572 | ||

Larp6
Genomic Map