siRNA Details
Gene Symbol: 4930524B15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026262.1 | NP_080538.1 | 1, 2 | 324 | ||
GenBank mRNA | AK015006.1 | - | 1, 2 | 366 | ||
AK015879.1 | - | 1, 2 | 324 | |||
BC114574.1 | - | 1, 2 | 314 | 40049349 |
