siRNA Details
| Gene Symbol: 4930519G04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026263.2 | NP_080539.1 | 9 | 797 | ||
| GenBank mRNA | AK005973.1 | - | 5 | 402 | ||
| AK015843.1 | - | 9 | 799 | |||
| AK015857.1 | - | 9 | 848 | |||
| AK161377.1 | - | 7 | 1698 | |||

4930519G04Rik
Genomic Map