siRNA Details
| Gene Symbol: Polr2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145632.2 | NP_663607.1 | 3 | 307 | ||
| GenBank mRNA | AK146429.1 | - | 3 | 332 | ||
| AK161617.1 | - | 3 | 328 | |||
| BC002306.1 | - | 3 | 307 | 3591968 | ||
| BC092087.1 | - | 3 | 217 | 5042770 | ||
| BC125612.1 | - | 3 | 291 | 40130127 | ||
| BC125638.1 | - | 3 | 291 | 40130153 | ||

Polr2h
Genomic Map