siRNA Details
| Gene Symbol: Lyrm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163628.1 | NP_001157100.1 | 2, 3 | 441 | ||
| NM_133688.3 | NP_598449.1 | 2, 3 | 204 | |||
| GenBank mRNA | AF412298.1 | - | 2, 3 | 858 | ||
| AF412299.1 | - | 3, 4 | 439 | |||
| AF412300.1 | - | 2, 3 | 205 | |||
| AK015530.1 | - | 2, 3 | 177 | |||
| AK075835.1 | - | 2, 3 | 185 | |||
| AY753325.1 | - | 2, 3 | 196 | |||
| BC021522.1 | - | 2, 3 | 152 | 5352723 | ||
| CX203242.1 | - | 2, 3 | 441 | |||

Lyrm5
Genomic Map