siRNA Details
| Gene Symbol: Dph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026344.3 | NP_080620.1 | 4 | 866 | ||
| GenBank mRNA | AK011199.1 | - | 4 | 884 | ||
| AK018638.1 | - | 4 | 867 | |||
| AK051026.1 | - | 4 | 886 | |||
| AK080747.1 | - | 3 | 1077 | |||
| BC014746.1 | - | 3 | 762 | |||
| BC031124.1 | - | 4 | 853 | 5029876 | ||

Dph2
Genomic Map