siRNA Details
| Gene Symbol: Rtp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023386.5 | NP_075875.3 | 2 | 419 | ||
| GenBank mRNA | AJ251364.1 | - | 2 | 414 | ||
| AK007477.1 | - | 2 | 397 | |||
| AK018021.1 | - | 2 | 412 | |||
| AY562228.1 | - | 2 | 349 | |||
| BC024872.1 | - | 2 | 377 | 3990099 | ||

Rtp4
Genomic Map