siRNA Details
| Gene Symbol: Ufm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026435.5 | NP_080711.1 | 3 | 148 | ||
| GenBank mRNA | AK007788.1 | - | 3 | 112 | ||
| AK008280.1 | - | 3 | 111 | |||
| AK011737.1 | - | 3 | 129 | |||
| AK089948.1 | - | 3 | 114 | |||
| AK148146.1 | - | 3 | 118 | |||
| AK166332.1 | - | 3 | 142 | |||
| AK167301.1 | - | 3 | 117 | |||
| AK167321.1 | - | 3 | 112 | |||
| BC051554.1 | - | 2 | 112 | |||
| BC061065.1 | - | 3 | 147 | 6594260 | ||
| CJ075089.1 | - | 3 | 174 | |||

Ufm1
Genomic Map