siRNA Details
| Gene Symbol: Coa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174987.4 | NP_778152.1 | 2 | 601 | ||
| GenBank mRNA | AK008041.1 | - | 2 | 536 | ||
| AK009404.1 | - | 2 | 575 | |||
| BC024399.1 | - | 2 | 531 | 5026081 | ||
| BC038634.1 | - | 2 | 587 | 1513986 | ||
| BC046907.1 | - | 3 | 492 | 6394880 | ||

Coa6
Genomic Map