siRNA Details
| Gene Symbol: Sumf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026445.2 | NP_080721.1 | 3, 4 | 364 | ||
| GenBank mRNA | AK013017.1 | - | 3, 4 | 344 | ||
| AK076022.1 | - | 3, 4 | 370 | |||
| AK138300.1 | - | 3, 4 | 355 | |||
| AK140169.1 | - | 3, 4 | 335 | |||
| AK157784.1 | - | 3, 4 | 365 | |||
| AK166216.1 | - | 3, 4 | 343 | |||
| BC104404.1 | - | 1, 2 | 473 | |||
| BC104405.1 | - | 1, 2 | 473 | |||
| BC119479.1 | - | 1, 2 | 473 | |||
| BC119480.2 | - | 1, 2 | 473 | 40050522 | ||

Sumf2
Genomic Map