siRNA Details
| Gene Symbol: Rnaseh2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026616.2 | NP_080892.1 | 4 | 545 | ||
| GenBank mRNA | AF346604.1 | - | 4 | 543 | ||
| AK011556.1 | - | 4 | 523 | |||
| AK018776.1 | - | 4 | 534 | |||
| BC024333.1 | - | 4 | 517 | 5040687 | ||
| CD553809.1 | - | 1 | 36 | |||

Rnaseh2c
Genomic Map