siRNA Details
| Gene Symbol: Sycn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026716.3 | NP_080992.3 | 1 | 143 | ||
| GenBank mRNA | AK003083.1 | - | 1 | 151 | ||
| BC019567.1 | - | 1 | 143 | 4208065 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sycn| Gene Symbol: Sycn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026716.3 | NP_080992.3 | 1 | 143 | ||
| GenBank mRNA | AK003083.1 | - | 1 | 151 | ||
| BC019567.1 | - | 1 | 143 | 4208065 | ||