siRNA Details
| Gene Symbol: Tomm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099675.1 | NP_001093145.1 | 2 | 336 | ||
| NM_001134646.1 | NP_001128118.1 | 3 | 386 | |||
| GenBank mRNA | AA423234.1 | - | 2 | 336 | ||
| AK003822.1 | - | 2 | 332 | |||
| CA466100.1 | - | 3 | 378 | |||

Tomm5
Genomic Map