siRNA Details
| Gene Symbol: Rnf122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175136.2 | NP_780345.1 | 6 | 1375 | ||
| GenBank mRNA | AK004356.1 | - | 1 | 594 | ||
| AK085376.1 | - | 6 | 1358 | |||
| AK171823.1 | - | 6 | 1400 | |||
| BC038399.1 | - | 6 | 1377 | 4947596 | ||
| BC050268.1 | - | 5 | 1065 | |||

Rnf122
Genomic Map