siRNA Details
| Gene Symbol: Prpf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133701.2 | NP_598462.1 | 20 | 2742 | ||
| GenBank mRNA | AK004462.1 | - | 5 | 441 | ||
| AK081998.1 | - | 20 | 2677 | |||
| AK145461.1 | - | 20 | 2745 | |||
| AK166890.1 | - | 20 | 2693 | |||
| AK219156.1 | - | 3 | 199 | |||
| BC005801.1 | - | 10 | 1258 | |||
| BC014869.1 | - | 20 | 2669 | 3602158 | ||
| BC023691.1 | - | 20 | 2671 | 5344332 | ||
| CJ228927.1 | - | 4 | 353 | |||

Prpf6
Genomic Map