siRNA Details
| Gene Symbol: Prpf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133701.2 | NP_598462.1 | 9 | 1301 | ||
| GenBank mRNA | AK081998.1 | - | 9 | 1236 | ||
| AK166890.1 | - | 9 | 1253 | |||
| BC014869.1 | - | 9 | 1228 | 3602158 | ||
| BC023691.1 | - | 9 | 1230 | 5344332 | ||
| BC025030.1 | - | 9 | 1247 | 4947416 | ||

Prpf6
Genomic Map