siRNA Details
| Gene Symbol: Zg16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026918.2 | NP_081194.1 | 3 | 400 | ||
| GenBank mRNA | BC031800.1 | - | 3 | 400 | 4208847 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zg16| Gene Symbol: Zg16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026918.2 | NP_081194.1 | 3 | 400 | ||
| GenBank mRNA | BC031800.1 | - | 3 | 400 | 4208847 | |