siRNA Details
| Gene Symbol: Tmem258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163431.1 | NP_001156903.1 | 3 | 318 | ||
| NM_026919.1 | NP_081195.1 | 3 | 289 | |||
| GenBank mRNA | AK007351.1 | - | 3 | 274 | ||
| AK145833.1 | - | 3 | 282 | |||
| AV456553.1 | - | 3 | 260 | |||
| BC058335.1 | - | 3 | 245 | |||
| BC085316.1 | - | 2 | 915 | |||

Tmem258
Genomic Map