siRNA Details
| Gene Symbol: 1810013L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081400.3 | NP_001074869.1 | 2, 3 | 674 | ||
| GenBank mRNA | AK007485.1 | - | 2, 3 | 674 | ||
| BC038008.1 | - | 1, 2 | 111 | |||
| BC116778.1 | - | 2, 3 | 657 | 40126097 | ||
| BC116780.1 | - | 2, 3 | 657 | 40126099 | ||

1810013L24Rik
Genomic Map