siRNA Details
| Gene Symbol: Sult1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026935.4 | NP_081211.3 | 7, 8 | 996 | ||
| GenBank mRNA | AK007384.1 | - | 7, 8 | 856 | ||
| AK144336.1 | - | 7, 8 | 936 | |||
| AY005469.1 | - | 7, 8 | 996 | |||
| BC022665.1 | - | 7, 8 | 989 | 4223678 | ||

Sult1c2
Genomic Map