siRNA Details
| Gene Symbol: Cd209f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026956.2 | NP_081232.2 | 8 | 710 | ||
| GenBank mRNA | BC021323.1 | - | 8 | 791 | ||
| BC038121.1 | - | 8 | 1113 | |||
| BC046292.1 | - | 6 | 751 | |||
| BC052782.1 | - | 8 | 701 | |||
| BC099497.1 | - | 8 | 705 | |||

Cd209f
Genomic Map