siRNA Details
| Gene Symbol: Ccdc23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038998.2 | NP_001034087.1 | 3 | 458 | ||
| NM_024462.2 | NP_077782.1 | 3 | 384 | |||
| GenBank mRNA | AI048353.1 | - | 4 | 336 | ||
| AK010418.1 | - | 3 | 314 | |||
| AK079783.1 | - | 3 | 243 | |||
| BC002274.1 | - | 3 | 297 | 3496493 | ||
| BF012097.1 | - | 3 | 388 | |||
| BQ044357.1 | - | 2 | 150 | |||
| CF105092.1 | - | 3 | 444 | |||

Ccdc23
Genomic Map