siRNA Details
| Gene Symbol: Tmem52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253856.1 | NP_001240785.1 | 5 | 506 | ||
| NM_001253858.1 | NP_001240787.1 | 4 | 464 | |||
| NM_027161.2 | NP_081437.1 | 5 | 524 | |||
| GenBank mRNA | AK009779.1 | - | 5 | 487 | ||
| BC144992.1 | - | 5 | 523 | 9053540 | ||
| BC150685.1 | - | 5 | 504 | 9087596 | ||

Tmem52
Genomic Map