siRNA Details
| Gene Symbol: Ndufaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027175.3 | NP_081451.3 | 2 | 657 | ||
| GenBank mRNA | AK010328.1 | - | 1 | 487 | ||
| AK050343.1 | - | 2 | 658 | |||
| BC018422.1 | - | 2 | 487 | 3593650 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufaf1| Gene Symbol: Ndufaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027175.3 | NP_081451.3 | 2 | 657 | ||
| GenBank mRNA | AK010328.1 | - | 1 | 487 | ||
| AK050343.1 | - | 2 | 658 | |||
| BC018422.1 | - | 2 | 487 | 3593650 | ||