siRNA Details
| Gene Symbol: Ndufaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027175.3 | NP_081451.3 | 5 | 1157 | ||
| GenBank mRNA | AK010328.1 | - | 4 | 987 | ||
| AK050343.1 | - | 5 | 1158 | |||
| BC018422.1 | - | 5 | 987 | 3593650 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufaf1| Gene Symbol: Ndufaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027175.3 | NP_081451.3 | 5 | 1157 | ||
| GenBank mRNA | AK010328.1 | - | 4 | 987 | ||
| AK050343.1 | - | 5 | 1158 | |||
| BC018422.1 | - | 5 | 987 | 3593650 | ||