siRNA Details
| Gene Symbol: Ipmk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027184.1 | NP_081460.1 | 6 | 768 | ||
| GenBank mRNA | AK036084.1 | - | 6 | 536 | ||
| AK172246.1 | - | 6 | 860 | |||
| BC052463.1 | - | 6 | 768 | 6834748 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ipmk| Gene Symbol: Ipmk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027184.1 | NP_081460.1 | 6 | 768 | ||
| GenBank mRNA | AK036084.1 | - | 6 | 536 | ||
| AK172246.1 | - | 6 | 860 | |||
| BC052463.1 | - | 6 | 768 | 6834748 | ||