siRNA Details
| Gene Symbol: 1700028J19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006462.1 | - | 5 | 1201 | ||
| BC049653.1 | - | 5 | 1105 | |||
| BC061239.1 | - | 4 | 1015 | |||
| BC087938.1 | - | 4 | 1015 | |||
| BC100477.1 | - | 5 | 1121 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700028J19Rik| Gene Symbol: 1700028J19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006462.1 | - | 5 | 1201 | ||
| BC049653.1 | - | 5 | 1105 | |||
| BC061239.1 | - | 4 | 1015 | |||
| BC087938.1 | - | 4 | 1015 | |||
| BC100477.1 | - | 5 | 1121 | |||