siRNA Details
| Gene Symbol: Cmtm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027294.2 | NP_081570.1 | 1 | 317 | ||
| GenBank mRNA | AK012262.1 | - | 1 | 318 | ||
| AK137729.1 | - | 1 | 323 | |||
| AK161079.1 | - | 1 | 327 | |||
| AY243513.1 | - | 1 | 352 | |||
| BC027554.1 | - | 1 | 239 | 1382818 | ||

Cmtm8
Genomic Map