siRNA Details
| Gene Symbol: 2210013O21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027327.1 | NP_081603.1 | 4 | 636 | ||
| GenBank mRNA | AK008724.1 | - | 4 | 602 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210013O21Rik| Gene Symbol: 2210013O21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027327.1 | NP_081603.1 | 4 | 636 | ||
| GenBank mRNA | AK008724.1 | - | 4 | 602 | ||