siRNA Details
| Gene Symbol: Fuz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027376.3 | NP_081652.2 | 11 | 1237 | ||
| GenBank mRNA | AK033247.1 | - | 11 | 1224 | ||
| AK134650.1 | - | 11 | 1237 | |||
| BC045601.1 | - | 8 | 2256 | |||
| BC057127.1 | - | 2 | 1784 | |||
| CK624547.1 | - | 6 | 499 | |||

Fuz
Genomic Map