siRNA Details
| Gene Symbol: Pak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027470.3 | NP_081746.1 | 7, 8 | 1657 | ||
| GenBank mRNA | AK017713.1 | - | 2, 3 | 281 | ||
| AK088512.1 | - | 7, 8 | 1634 | |||
| AK129298.2 | - | 7, 8 | 1653 | |||
| AY217016.1 | - | 5, 6 | 1372 | |||
| BC030389.1 | - | 4, 5 | 815 | |||
| BC048238.1 | - | 7, 8 | 1619 | 6475114 | ||

Pak4
Genomic Map