siRNA Details
| Gene Symbol: Gulp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028450.3 | NP_082726.2 | 6, 7 | 644 | ||
| GenBank mRNA | AK014093.1 | - | 5, 6 | 393 | ||
| AK017798.1 | - | 6, 7 | 622 | |||
| AK028490.1 | - | 5, 6 | 560 | |||
| AK036670.1 | - | 6, 7 | 644 | |||
| BC032154.1 | - | 6, 7 | 815 | 5068339 | ||
| BY713087.1 | - | 5, 6 | 391 | |||
| DQ786776.1 | - | 5, 6 | 398 | |||

Gulp1
Genomic Map