siRNA Details
| Gene Symbol: Rasl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033158.2 | NP_001028330.2 | 5 | 1779 | ||
| GenBank mRNA | AK014511.1 | - | 5 | 1742 | ||
| AK171623.1 | - | 5 | 1735 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasl12| Gene Symbol: Rasl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033158.2 | NP_001028330.2 | 5 | 1779 | ||
| GenBank mRNA | AK014511.1 | - | 5 | 1742 | ||
| AK171623.1 | - | 5 | 1735 | |||