siRNA Details
| Gene Symbol: Clec2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168223.1 | NP_001161695.1 | 2 | 56 | ||
| NM_001168224.1 | NP_001161696.1 | 2 | 168 | |||
| NM_027562.4 | NP_081838.1 | 3 | 230 | |||
| GenBank mRNA | AK014570.1 | - | 3 | 231 | ||
| AK137753.1 | - | 2 | 166 | |||
| AY137341.1 | - | 2 | 168 | |||
| AY137342.1 | - | 2 | 56 | |||
| BC117543.1 | - | 3 | 220 | 40105656 | ||
| JF792238.1 | - | 2 | 67 | |||
| JF792239.1 | - | 2 | 118 | |||

Clec2g
Genomic Map